Learn More
Sino Biological Anti-CD79A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100128RP02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CD79a, and purified by antigen affinity chromatography.
Catalog No. 50-161-0499