Learn More
Sino Biological Anti-Cytokeratin 14/KRT14 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 100129MM02T100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Catalog No. 50-161-0505