Learn More
Sino Biological Anti-Cytokeratin 14/KRT14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100129RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the C-terminus of the human Cytokeratin 14, and purified by antigen affinity chromatography.
Catalog No. 50-161-0510