Learn More
Sino Biological Recombinant Human FGF1 Protein
Supplier:  Sino Biological 10013HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human FGF acidic (AAA79245.1) (Phe 16-Asp 155) was expressed, with an additional Met at the N-terminus.
Catalog No. 50-161-3803