Learn More
Sino Biological Recombinant Anti-Olig2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100137R064100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the N-terminus of the Human Olig-2.
Catalog No. 50-161-0575