Learn More
Sino Biological Recombinant Anti-GFAP Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100140R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human GFAP.
Catalog No. 50-161-0598