Learn More
Sino Biological Recombinant Human GM-CSF/CSF2 Protein (His Tag)
Supplier:  Sino Biological 10015H07H1
Encompass_Preferred
A DNA sequence encoding the mature form of human GM-CSF (NP_000749.2) (Ala 18-Glu 144) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-3814