Learn More
Sino Biological Recombinant Human GM-CSF/CSF2 Protein, HPLC-verified
Supplier:  Sino Biological 10015HNAH100
Encompass_Preferred
A DNA sequence encoding human GMCSF (NP_000749.2) (Met1-Glu144) was expressed.
Catalog No. 50-161-3819