Learn More
Sino Biological Recombinant Human GM-CSF/CSF2 Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 10015HNAHB20
Encompass_Preferred
A DNA sequence encoding the human CSF2 (NP_000749.2) (Met1-Glu144) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-161-3822