Learn More
Sino Biological MERS-CoV Spike Protein S1 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 100207RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the center region of the S1 subunit of MERS-CoV (NCoV / Novel coronavirus) spike glycoprotein (S protein), and purified by antigen affinity chromatography.
Catalog No. 50-161-0638