Learn More
Sino Biological Recombinant Human 4EBP1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10022H07E100
Encompass_Preferred
A DNA sequence encoding the human 4EBP1 (NP_004086.1) (Ser 2-Ile 118) with a N-terminal polyhistidine tag was expressed.
Catalog No. 50-161-3837