Learn More
Sino Biological Anti-4EBP1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10022MM04F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 4E-BP1 / EIF4EBP1 (rh 4E-BP1 / EIF4EBP1; Catalog#10022-H07E; NP_004086.1; Ser2-Ile118) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-0671