Learn More
Sino Biological Recombinant Anti-4EBP1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10022R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human 4E-BP1 / EIF4EBP1 (rh 4E-BP1 / EIF4EBP1; Catalog#10022-H07E; NP_004086.1; Ser2-Ile118).
Catalog No. 50-161-0677