Learn More
Sino Biological Anti-Connexin 43/GJA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100246T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human GJA1 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0714