Learn More
Sino Biological Anti-Chromogranin A/CHGA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100256T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human Chromogranin A/CHGA/CGA, and purified by antigen affinity chromatography.
Catalog No. 50-161-0738