Learn More
Sino Biological Anti-GSTP1/GST3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100259T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human GSTπ(GlutathioneS-Transferase,pi), and purified by antigen affinity chromatography.
Catalog No. 50-161-0746