Learn More
Sino Biological Anti-Enolase 2/ENO2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100261T40100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human NSE(Gamma-enolase) , and purified by antigen affinity chromatography.
Catalog No. 50-161-0752