Learn More
Sino Biological Anti-p63 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100263T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human p63(delta P63 /ΔN P63), and purified by antigen affinity chromatography.
Catalog No. 50-161-0755