Learn More
Sino Biological Recombinant Anti-p63 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100266R143100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with A synthetic peptide corresponding to the C-terminus of the human p63(alpha p63).
Catalog No. 50-161-0758