Learn More
Sino Biological Anti-Androgen receptor Antibody, Rabbit Polyclonal
Catalog No. 501610766
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the human Androgen receptor . Human Androgen receptor specific IgG was purified by Human Androgen receptor affinity chromatography.
Catalog No. 50-161-0766 Supplier Sino Biological Supplier No. 100272T32100
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the human Androgen receptor . Human Androgen receptor specific IgG was purified by Human Androgen receptor affinity chromatography.