Learn More
Sino Biological Anti-Androgen receptor Antibody, Rabbit Polyclonal
Catalog No. 501610769
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Androgen receptor / AR, and purified by antigen affinity chromatography.
Catalog No. 50-161-0769 Supplier Sino Biological Supplier No. 100273T32100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Androgen receptor / AR, and purified by antigen affinity chromatography.