Learn More
Sino Biological Anti-Synaptophysin/SYP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100298T40100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human Synaptophysin , and purified by antigen affinity chromatography.
Catalog No. 50-161-0796