Learn More
Sino Biological Anti-BMI1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100299T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human BMI1, and purified by antigen affinity chromatography.
Catalog No. 50-161-0801