Learn More
Sino Biological Recombinant Human ERK2 Protein (GST Tag), HPLC-verified
Supplier:  Sino Biological 10030H09B50
Encompass_Preferred
A DNA sequence encoding the human ERK2 (NP_002736.3) (Met 1-Ser 360) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-3842