Learn More
Sino Biological Anti-ERK2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10030MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ERK2 / MAPK1 / MAPK2 (rh ERK2 / MAPK1 / MAPK2; Catalog#10030-H09B; NP_002736.3; Met1-Ser360). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0836