Learn More
Sino Biological Anti-ERK2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10030T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ERK2/MAPK1/MAPK2 (rh ERK2/MAPK1/MAPK2; Catalog#10030-H09B; NP_002736.3; Met1-Ser360). ERK2/MAPK1/MAPK2 specific IgG was purified by Human ERK2/MAPK1/MAPK2 affinity chromatography.
Catalog No. 50-161-0846