Learn More
Sino Biological Anti-Cytokeratin 7/KRT7 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 100310MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the center region of the Human KRT7 / Cytokeratin-7. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0849