Learn More
Sino Biological Anti-Cytokeratin 7/KRT7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100311T32100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human KRT7 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0855