Learn More
Sino Biological Recombinant Anti-Cytokeratin 5/KRT5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100312R176100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Cytokeratin 5/KRT5.
Catalog No. 50-161-0864