Learn More
Sino Biological Anti-PIP/GCDFP 15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100315T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PIP/GCDFP-15, and purified by antigen affinity chromatography.
Catalog No. 50-161-0880