Learn More
Sino Biological Anti-IL-12 p35/IL12A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100321T32100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the mouse IL12A . Mouse IL12A specific IgG was purified by Mouse IL12A affinity chromatography.
Catalog No. 50-161-0892