Learn More
Sino Biological Anti-IL-15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100322T32100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the mouse mIL15 , and purified by antigen affinity chromatography.
Catalog No. 50-161-0895