Learn More
Sino Biological Anti-Noggin/NOG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100339T36100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human NOG, and purified by antigen affinity chromatography.
Catalog No. 50-161-0907