Learn More
Sino Biological Recombinant Anti-CD45 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100342R101100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the C-terminus of the Human CD45/PTPRC.
Catalog No. 50-161-0919