Learn More
Sino Biological Anti-Peripherin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100361T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PRPH / Peripherin, and purified by antigen affinity chromatography.
Catalog No. 50-161-0971