Learn More
Sino Biological Anti-CEBP alpha/CEBPA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100364T36100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the C-terminus of the human CEBPA . Huamn CEBPA specific IgG was purified by Huamn CEBPA affinity chromatography.
Catalog No. 50-161-0977