Learn More
Sino Biological Anti-V5 tag Antibody, Mouse Monoclonal
Supplier:  Sino Biological 100378MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the V5 tag sequence (GKPIPNPLLGLDST). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0996