Learn More
Sino Biological Anti-V5 tag Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100378T36100
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the V5-tag sequence. V5-tag specific IgG was purified by V5-taq affinity chromatography.
Catalog No. 50-161-0999