Learn More
Sino Biological Anti-ER alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100386T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human ER alpha/ESR1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1005