Learn More
Sino Biological Recombinant Human CD27/TNFRSF7 Protein (rFc Tag), HPLC-verified
Supplier:  Sino Biological 10039H31H200
Encompass_Preferred
A DNA sequence encoding the human CD27 (P26842) (Met1-Ile192) was expressed, fused with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-161-3849