Learn More
Sino Biological Anti-CD27/TNFRSF7 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10039MM09F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD27 / TNFRSF7 (rh CD27 / TNFRSF7; Catalog#10039-H03H; NP_001233.1; Met 1-Ile 192) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-1051