Learn More
Sino Biological Recombinant Anti-CD27/TNFRSF7 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10039R023100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD27 (rh CD27; Catalog#10039-H03H; NP_001233.1; Met1-Ile192).
Catalog No. 50-161-1054