Learn More
Sino Biological Recombinant Anti-CD27/TNFRSF7 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10039R025P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD27 (rh CD27; Catalog#10039-H03H; Met 1-Ile 192; NP_001233.1) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1072