Learn More
Sino Biological Anti-CD10/Neprilysin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100390T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CD10 / Neprilysin / MME, and purified by antigen affinity chromatography.
Catalog No. 50-161-1011