Learn More
Sino Biological Recombinant Anti-Glypican 3/GPC3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100393R024100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Glypican 3 / GPC3 / OCI-5. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1020