Learn More
Sino Biological Recombinant Anti-Glypican 3/GPC3 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 100393R024P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Glypican 3 / GPC3 / OCI-5, and it was conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1027