Learn More
Sino Biological Anti-Integrin alpha 3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100397T08100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human Integrin alpha 3/ITGA3/CD49C, and purified by antigen affinity chromatography.
Catalog No. 50-161-1037