Learn More
Sino Biological Recombinant Human CD30 Ligand/TNFSF8 Protein
Supplier:  Sino Biological 10040HNCH200
Encompass_Preferred
A DNA sequence encoding the human CD30L (NP_001235.1) (Gln63-Asp234) was expressed with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-3853