Learn More
Sino Biological Anti-CXCR3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100409T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CXCR3/CD183, and purified by antigen affinity chromatography.
Catalog No. 50-161-1099