Learn More
Sino Biological Recombinant Human 4-1BB/CD137 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10041H08H100
Encompass_Preferred
A DNA sequence encoding the human 4-1BB extracellular domain (NP_001552.2) (Met 1-Gln 186) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3858